Learn More
Crescent Chemical Co Inc PUROMYCIN-2HCL RESEARCH GRADE
SDP
Catalog No. 50248081
Encompass_Preferred
Puromycin-2HCl research grade From Streptomyces albo-niger. Free base (Mr 471.5) 86 %. Inhibitor of protein biosynthesis. Causes premature termination of the nascent polypeptide chain by its action as aminoacyl-tRNA analog (in procaryotes and eucaryotes). S
Catalog No. 50-248-081 Supplier Crescent Chemical Co Inc Supplier No. SE33835.01
Puromycin-2HCl research grade From Streptomyces albo-niger. Free base (Mr 471.5) 86 %. Inhibitor of protein biosynthesis. Causes premature termination of the nascent polypeptide chain by its action as aminoacyl-tRNA analog (in procaryotes and eucaryotes). S