Learn More
Sigma Aldrich Fine Chemicals Biosciences Puromycin dihydrochloride from Streptomyces alboniger, 58-58-2, MFCD00012691, 25 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P883325MG
Encompass
Empirical Formula (Hill Notation): C22H29N7O5 · 2HCl, Molecular Weight: 544.43, Synonym: 3′-[α-Amino-p-methoxyhydrocinnamamido]-3′-deoxy-N,N-dimethyladenosine dihydrochloride. Powder, BioReagent, suitable for cell culture. Puromycin dihydrochloride from Streptomyces alboniger has been used: in the preparation of puromycin stock solution for puromycin sensitivity assay, and to select lentivirus-infected cells containing puromycin resistant selection marker.
Catalog No. NC0734808