Learn More
Invivogen PUROMYCIN-500MG
SDP
Catalog No. NC1999692
Encompass
Puromycin, 500MG, CAS# 58-58-2, Puromycin is an aminonucleoside antibiotic produced by Streptomyces alboniger. It specifically inhibits peptidyl transfer on both prokaryotic and eukaryotic ribosomes. This antibiotic inhibits the growth of Gram positive bacteria and various animal and insect cells.
Catalog No. NC1999692 Supplier Invivogen Supplier No. ANTPR5B
Puromycin, 500MG, CAS# 58-58-2, Puromycin is an aminonucleoside antibiotic produced by Streptomyces alboniger. It specifically inhibits peptidyl transfer on both prokaryotic and eukaryotic ribosomes. This antibiotic inhibits the growth of Gram positive bacteria and various animal and insect cells.