Learn More
Invivogen Puromycin, 500 mg
SDP
Catalog No. NC9138918
Encompass
Puromycin is an aminonucleoside antibiotic produced by Streptomyces alboniger. It specifically inhibits peptidyl transfer on both prokaryotic and eukaryotic ribosomes. This antibiotic inhibits the growth of Gram positive bacteria and various animal and insect cells.
Catalog No. NC9138918 Supplier Invivogen Supplier No. ANTPR5
Puromycin is an aminonucleoside antibiotic produced by Streptomyces alboniger. It specifically inhibits peptidyl transfer on both prokaryotic and eukaryotic ribosomes. This antibiotic inhibits the growth of Gram positive bacteria and various animal and insect cells.