Learn More
Medchemexpress LLC HY-15695 500mg Medchemexpress, Puromycin aminonucleoside CAS:58-60-6 Purity:>98%
SDP
Catalog No. 502029159
Encompass_Preferred
Medchemexpress, HY-15695 500mg Puromycin aminonucleoside CAS:58-60-6 Puromycin aminonucleoside (NSC 3056) is the aminonucleoside portion of the antibiotic puromycin, and used in nephrosis animal models. Puromycin aminonucleoside induces apoptosis[2]. Puromycin aminonucleoside is a reversible inhibitor of dipeptidyl peptidase II and cytosol alanyl aminopeptidase[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9159 Supplier Medchemexpress LLC Supplier No. HY15695500MG
Medchemexpress, HY-15695 500mg Puromycin aminonucleoside CAS:58-60-6 Puromycin aminonucleoside (NSC 3056) is the aminonucleoside portion of the antibiotic puromycin, and used in nephrosis animal models. Puromycin aminonucleoside induces apoptosis[2]. Puromycin aminonucleoside is a reversible inhibitor of dipeptidyl peptidase II and cytosol alanyl aminopeptidase[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.