Learn More
Cayman Chemical PuromycIn AmInonucleosIde 10mg
SDP
Catalog No. 502969535
Encompass_Preferred
The aminonucleoside portion of PANS that is used to induce nephropathy in laboratory animal models to study focal and segmental glomerulosclerosis, the induction of nephrosis, and the events leading to proteinuria in rats
Catalog No. 50-296-9535 Supplier Cayman Chemical Supplier No. 1550910 MG
The aminonucleoside portion of PANS that is used to induce nephropathy in laboratory animal models to study focal and segmental glomerulosclerosis, the induction of nephrosis, and the events leading to proteinuria in rats