Learn More
Cayman Chemical PuromycIn AmInonucleosIde 25mg
SDP
Catalog No. 502971279
Encompass_Preferred
The aminonucleoside portion of PANS that is used to induce nephropathy in laboratory animal models to study focal and segmental glomerulosclerosis, the induction of nephrosis, and the events leading to proteinuria in rats
Catalog No. 50-297-1279 Supplier Cayman Chemical Supplier No. 1550925 MG
The aminonucleoside portion of PANS that is used to induce nephropathy in laboratory animal models to study focal and segmental glomerulosclerosis, the induction of nephrosis, and the events leading to proteinuria in rats