Learn More
Cayman Chemical Puromycin aminonucleoside 50MG
SDP
Catalog No. 502969470
Encompass_Preferred
The aminonucleoside portion of PANS that is used to induce nephropathy in laboratory animal models to study focal and segmental glomerulosclerosis, the induction of nephrosis, and the events leading to proteinuria in rats
Catalog No. 50-296-9470 Supplier Cayman Chemical Supplier No. 1550950 MG
The aminonucleoside portion of PANS that is used to induce nephropathy in laboratory animal models to study focal and segmental glomerulosclerosis, the induction of nephrosis, and the events leading to proteinuria in rats