Learn More
Enzo Life Sciences Puromycin Aminonucleoside (50mg). CAS: 58-60-6
SDP
Catalog No. 502010237
Encompass_Preferred
A puromycin analog which does not inhibit protein synthesis or induce apoptosis. It is a glomerular epithelial cell toxin which may be used to induce nephropathy in laboratory animals. Alternative name: 3'-Amino-3'-Deoxy-N,N-dimethyladenosine. Purity: ≥98% (TLC). Solubility: Soluble in water (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0237 Supplier Enzo Life Sciences Supplier No. BMLA2600050
A puromycin analog which does not inhibit protein synthesis or induce apoptosis. It is a glomerular epithelial cell toxin which may be used to induce nephropathy in laboratory animals. Alternative name: 3'-Amino-3'-Deoxy-N,N-dimethyladenosine. Purity: ≥98% (TLC). Solubility: Soluble in water (10mg/ml). Long Term Storage: -20°C.