Learn More
Medchemexpress LLC HY-15695 100mg Medchemexpress, Puromycin aminonucleoside 3'-Amino-3'-deoxy-N6,N6-dimethyladenosine CAS:58-60-6 Purity:98%
SDP
Catalog No. 501638381
Encompass_Preferred
Medchemexpress,HY-15695 100mg Puromycin aminonucleoside 3'-Amino-3'-deoxy-N6,N6-dimethyladenosine CAS:58-60-6 Formula:C12H18N6O3 Puromycin aminonucleoside is the aminonucleoside portion of the antibiotic puromycin, and a puromycin analog which does not inhibit protein synthesis or induce apoptosis. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8381 Supplier Medchemexpress LLC Supplier No. HY15695100MG
Medchemexpress,HY-15695 100mg Puromycin aminonucleoside 3'-Amino-3'-deoxy-N6,N6-dimethyladenosine CAS:58-60-6 Formula:C12H18N6O3 Puromycin aminonucleoside is the aminonucleoside portion of the antibiotic puromycin, and a puromycin analog which does not inhibit protein synthesis or induce apoptosis. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.