Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Puromycin-d3 | 98.2% | 10 MG
SDP

Catalog No. 5000297823
Encompass_Preferred

Puromycin-d3 is a deuterium-labeled analogue of puromycin used as a labeled standard and tracer in biochemical research and mass spectrometry applications. The deuterated isotopologue enables accurate quantitation and tracking in proteomics and metabolism studies.

  • Deuterium-labeled puromycin (d3) for use as an internal standard in mass spectrometry and tracer studies.
  • Aminoglycoside antibiotic that inhibits protein synthesis, useful for translation and inhibitor assays.
  • Molecular formula C22H26D3N7O5; molecular weight 474.53 g/mol.
  • Purity 98.2% as supplied by the manufacturer.
  • Supplied as a powder, typically in 10 mg vials; store at -20°C for long-term stability.

Catalog No. 50-002-97823 Supplier Medchemexpress LLC Supplier No. HYB1743S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puromycin-d3 is a deuterium-labeled analogue of puromycin used as a labeled standard and tracer in biochemical research and mass spectrometry applications. The deuterated isotopologue enables accurate quantitation and tracking in proteomics and metabolism studies.

  • Deuterium-labeled puromycin (d3) for use as an internal standard in mass spectrometry and tracer studies.
  • Aminoglycoside antibiotic that inhibits protein synthesis, useful for translation and inhibitor assays.
  • Molecular formula C22H26D3N7O5; molecular weight 474.53 g/mol.
  • Purity 98.2% as supplied by the manufacturer.
  • Supplied as a powder, typically in 10 mg vials; store at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.