Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Puromycin-d3 dihydrochloride | 1 MG
SDP

Catalog No. 5000413476
Encompass_Preferred

Puromycin-d3 dihydrochloride is a deuterium-labeled aminonucleoside antibiotic that inhibits protein synthesis. Stable heavy isotopes have been incorporated into this drug molecule for use as a tracer in drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.

  • Inhibits protein synthesis.
  • Deuterium labeled compound.
  • Functions as a tracer for quantitation in drug development.
  • Affects pharmacokinetic and metabolic profiles.
  • Molecular formula: C₂₂H₂₈D₃Cl₂N₇O₅
  • Molecular weight: 547.45 g/mol
  • Targets bacterial, antibiotic, and isotope-labeled compounds.
  • Soluble in methanol, DMSO, and ethanol.

Catalog No. 50-004-13476 Supplier Medchemexpress LLC Supplier No. HYB1743AS1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puromycin-d3 dihydrochloride is a deuterium-labeled aminonucleoside antibiotic that inhibits protein synthesis. Stable heavy isotopes have been incorporated into this drug molecule for use as a tracer in drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.

  • Inhibits protein synthesis.
  • Deuterium labeled compound.
  • Functions as a tracer for quantitation in drug development.
  • Affects pharmacokinetic and metabolic profiles.
  • Molecular formula: C₂₂H₂₈D₃Cl₂N₇O₅
  • Molecular weight: 547.45 g/mol
  • Targets bacterial, antibiotic, and isotope-labeled compounds.
  • Soluble in methanol, DMSO, and ethanol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.