Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Puromycin Dihydrochloride

Catalog No. MT61385RA Shop All Corning Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT61385RA 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT61385RA Supplier Corning™ Supplier No. 61385RA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Packaged in plastic bottles

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -5° to -25°C
Form Powder
Product Type Antibiotic
Format Plastic Bottle
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.