Learn More
Research Products International Corp Puromycin Dihydrochloride, 25 Milligrams
SDP
Catalog No. 50488917
Encompass_Preferred
Puromycin Dihydrochloride is a potent antibiotic. It is an amino nucleoside antibiotic that mimics the 3'-end of aminoacyl-tRNA. It enters the ribosomal A site during translation, causing premature chain termination and inhibiting protein synthesis. Puromycin is incorporated into growing polypeptide chains, leading to the release of incomplete and nonfunctional peptides. 
Widely used as a tool in cell biology to study protein synthesis and investigate translation processes. Its ability to terminate protein synthesis at the ribosome makes it valuable for various applications, including pulse-chase experiments and the analysis of nascent peptides. Puromycin resistance genes are often used as selectable markers in genetic engineering. Cells expressing the resistance gene can survive and proliferate in the presence of Puromycin, allowing for the selection of transfected or transduced cells.
Catalog No. 50-488-917 Supplier Research Products International Corp Supplier No. P330200.025
Puromycin Dihydrochloride is a potent antibiotic. It is an amino nucleoside antibiotic that mimics the 3'-end of aminoacyl-tRNA. It enters the ribosomal A site during translation, causing premature chain termination and inhibiting protein synthesis. Puromycin is incorporated into growing polypeptide chains, leading to the release of incomplete and nonfunctional peptides. 
Widely used as a tool in cell biology to study protein synthesis and investigate translation processes. Its ability to terminate protein synthesis at the ribosome makes it valuable for various applications, including pulse-chase experiments and the analysis of nascent peptides. Puromycin resistance genes are often used as selectable markers in genetic engineering. Cells expressing the resistance gene can survive and proliferate in the presence of Puromycin, allowing for the selection of transfected or transduced cells.