Learn More
Gold Biotechnology Inc Puromycin Dihydrochloride 1 g
SDP
Catalog No. 501532828
Encompass_Preferred
Puromycin is an aminonucleoside antibiotic that causes early chain termination within the ribosome during translation. This inhibition of translation is effective in both in vivo and in vitro applications, making it an ideal tool for protein synthesis studies. While selection for E. coli cells is possible with puromycin, it is more commonly and effectively used for mammalian cell selection.
Catalog No. 50-153-2828 Supplier Gold Biotechnology Inc Supplier No. P6001
Puromycin is an aminonucleoside antibiotic that causes early chain termination within the ribosome during translation. This inhibition of translation is effective in both in vivo and in vitro applications, making it an ideal tool for protein synthesis studies. While selection for E. coli cells is possible with puromycin, it is more commonly and effectively used for mammalian cell selection.