Learn More
Sigma Aldrich Fine Chemicals Biosciences Puromycin dihydrochloride, 58-58-2, MFCD00012691, 10mL
Catalog No. 501657050
Encompass
Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, 0.2 μm filtered
Catalog No. 50-165-7050 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P962010ML
Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, 0.2 μm filtered