Learn More
Medchemexpress LLC Puromycin Dihydrochloride | 58-58-2 | MFCD00012691 | ≥98% | 544.43 | 5 ML
SDP
Catalog No. 5000430571
Encompass_Preferred
Puromycin is an aminonucleoside antibiotic produced by *Streptomyces alboniger*. It inhibits protein synthesis by disrupting peptide transfer on ribosomes, causing premature chain termination during translation. This antibiotic is capable of killing most gram-positive bacteria and various animal or insect cells.
  • Inhibits protein synthesis.
  • Disrupts peptide transfer on ribosomes.
  • Causes premature chain termination during translation.
  • Kills most gram-positive bacteria.
  • Kills various animal or insect cells.
  • Effective for selecting vectors bearing the pac gene in mammalian cell lines.
  • Effective for selecting vectors bearing the pac gene in E. coli.
Catalog No. 50-004-30571 Supplier Medchemexpress LLC Supplier No. HYK10575ML
Puromycin is an aminonucleoside antibiotic produced by *Streptomyces alboniger*. It inhibits protein synthesis by disrupting peptide transfer on ribosomes, causing premature chain termination during translation. This antibiotic is capable of killing most gram-positive bacteria and various animal or insect cells.
  • Inhibits protein synthesis.
  • Disrupts peptide transfer on ribosomes.
  • Causes premature chain termination during translation.
  • Kills most gram-positive bacteria.
  • Kills various animal or insect cells.
  • Effective for selecting vectors bearing the pac gene in mammalian cell lines.
  • Effective for selecting vectors bearing the pac gene in E. coli.