Learn More
Medchemexpress LLC Puromycin dihydrochloride | 58-58-2 | MFCD00012691 | 99.9% | 500 MG
SDP
Catalog No. 5000413475
Encompass_Preferred
Puromycin dihydrochloride is an aminonucleoside antibiotic that inhibits protein synthesis. It blocks protein synthesis after aminoacyl-sRNA formation, leading to the accumulation of small peptides. It induces premature termination of translation by serving as an acceptor substrate and by competing with aminoacyl-tRNA for binding to the A' site. This allows for direct evaluation of translation activity in single cells using immunofluorescence microscopy and in heterogeneous cell populations via fluorescence-activated cell sorting.
  • Aminonucleoside antibiotic
  • Inhibits protein synthesis
  • Blocks protein synthesis after aminoacyl-sRNA formation
  • Leads to the accumulation of small peptides
  • Causes premature termination of translation
  • Allows direct evaluation of translation activity in single cells by immunofluorescence microscopy and in heterogeneous cell populations by fluorescence-activated cell sorting
Catalog No. 50-004-13475 Supplier Medchemexpress LLC Supplier No. HYB1743A500MG
Puromycin dihydrochloride is an aminonucleoside antibiotic that inhibits protein synthesis. It blocks protein synthesis after aminoacyl-sRNA formation, leading to the accumulation of small peptides. It induces premature termination of translation by serving as an acceptor substrate and by competing with aminoacyl-tRNA for binding to the A' site. This allows for direct evaluation of translation activity in single cells using immunofluorescence microscopy and in heterogeneous cell populations via fluorescence-activated cell sorting.
  • Aminonucleoside antibiotic
  • Inhibits protein synthesis
  • Blocks protein synthesis after aminoacyl-sRNA formation
  • Leads to the accumulation of small peptides
  • Causes premature termination of translation
  • Allows direct evaluation of translation activity in single cells by immunofluorescence microscopy and in heterogeneous cell populations by fluorescence-activated cell sorting