Learn More
Sigma Aldrich Fine Chemicals Biosciences Puromycin dihydrochloride from Streptomyces alboniger, 58-58-2, MFCD00012691, 10mg
Catalog No. 501657049
Encompass
Empirical Formula (Hill Notation) C22H29N7O5 · 2HCl, Molecular Weight 544.43, powder, BioReagent, suitable for cell culture, Synonym: 3′-[α-Amino-p-methoxyhydrocinnamamido]-3′-deoxy-N,N-dimethyladenosine dihydrochloride
Catalog No. 50-165-7049 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P883310MG
Empirical Formula (Hill Notation) C22H29N7O5 · 2HCl, Molecular Weight 544.43, powder, BioReagent, suitable for cell culture, Synonym: 3′-[α-Amino-p-methoxyhydrocinnamamido]-3′-deoxy-N,N-dimethyladenosine dihydrochloride