Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Puromycin dihydrochloride hydrate, 99%

Catalog No. AC227420500
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227420500 50 mg Glass bottle
AC227420100 10 mg Glass bottle
AC227422500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC227420500 Supplier Thermo Scientific Chemicals Supplier No. 227420500
  • $253.00
    $203.65 / Each of 1

    Save $49.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puromycin dihydrochloride hydrate, CAS # 58-58-2, is an aminoglycoside showing antibiotic, anti-neoplastic, and anti-protozoal activities. This compound also has the ability to inhibit protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Puromycin dihydrochloride hydrate is the hydrochloride salt form of puromycin and is an aminoglycoside with antibiotic activity isolated from the Streptomyces alboniger
• Puromycin dihydrochloride hydrate is a protein synthesis inhibitor that causes premature chain termination by acting as an analog of the 3’-terminal ends of aminoacyl-tRNA, inhibiting protein synthesis

Applications

• This compound shows anti-microbial, anti-protozoal, anti-trypanosomal, and anti-neoplastic activities and is also used as an antibiotic in cell culture
• It also reveals anti-tumor activity in studies on brain cancer cells
• This compound has been used to study transcriptional regulatory mechanisms that control the sequential and coordinate expression of genes during cell differentiation
• It is active against gram-positive microorganisms, less active against acid-fast bacilli and gram-negative microorganisms. Puromycin can prevent growth of bacteria, protozoa, algae, and mammalian cells in in vitro studies.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-58-2
Molecular Formula C22H31Cl2N7O5
Molecular Weight (g/mol) 544.43
MDL Number MFCD00150080
InChI Key MKSVFGKWZLUTTO-MLYJQVMKNA-N
Synonym puromycin dihydrochloride
PubChem CID 131632508
IUPAC Name (2S)-2-amino-N-[(2S,3S,4S,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride
SMILES Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1

Specifications

Melting Point 168.0°C to 178.0°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 50 mg
Beilstein 26,IV,3704
Merck Index 15,8054
Solubility Information Solubility in water: soluble. Other solubilities: less soluble in ethanol
Formula Weight 544.44
Percent Purity 99%
Chemical Name or Material Puromycin dihydrochloride hydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
EINECSNumber 200-387-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.