Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puromycin dihydrochloride hydrate, 99%, Thermo Scientific™

Catalog No. AC227420250
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC227420250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC227420250 Supplier Thermo Scientific Chemicals Supplier No. 227420250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-58-2
Molecular Formula C22H31Cl2N7O5
Molecular Weight (g/mol) 544.43
MDL Number MFCD00150080
InChI Key MKSVFGKWZLUTTO-MLYJQVMKNA-N
Synonym puromycin dihydrochloride
PubChem CID 131632508
IUPAC Name (2S)-2-amino-N-[(2S,3S,4R,5R)-5-[6-(dimethylamino)-9H-purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide dihydrochloride
SMILES Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1

Specifications

Melting Point 168.0°C to 178.0°C
Quantity 25 mg
Solubility Information Solubility in water: soluble. Other solubilities: less soluble in ethanol
Formula Weight 544.44
Chemical Name or Material Puromycin dihydrochloride hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.