Learn More
Abcam Puromycin dihydrochloride, Protein biosynthesis inhibitor, 25MG
Catalog No. 502706600
Encompass_Preferred
MW 544.4 Da, Purity >98%. Aminonuclease antibiotic agent. Protein biosynthesis inhibitor. Induces cell cycle arrest in G2/M phase and apoptosis. Inhibits transport of proteins into mitochondria. Inhibits translation in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6600 Supplier Abcam Supplier No. AB14145325MG
Add to Cart
Add to Cart
MW 544.4 Da, Purity >98%. Aminonuclease antibiotic agent. Protein biosynthesis inhibitor. Induces cell cycle arrest in G2/M phase and apoptosis. Inhibits transport of proteins into mitochondria. Inhibits translation in vivo.
The product is subject to the following: Abcam Restricted Use Statement