Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puromycin Dihydrochloride, TCI America™
SDP

Catalog No. P2311100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2311100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2311100MG Supplier TCI America Supplier No. P2311100MG

missing translation for 'validMainframeMsg'

Specifications

Type Puromycin Dihydrochloride
Physical Form Crystalline Powder
CAS 58-58-2
Melting Point 174°C
Quantity 100 mg
Molecular Formula C22H31Cl2N7O5
MDL Number MFCD00150080
InChI Key MKSVFGKWZLUTTO-MLYJQVMKNA-N
SMILES Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1
IUPAC Name (2S)-2-amino-N-[(2S,3S,4R,5R)-5-[6-(dimethylamino)-9H-purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide dihydrochloride
Molecular Weight (g/mol) 544.43
PubChem CID 131632508
Formula Weight 544.43
Percent Purity ≥97.0% (HPLC)
Chemical Name or Material Puromycin Dihydrochloride
Show More Show Less
RTECSNumber AU7355000
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.