Learn More
ABclonal Technology Puromycin Rabbit mAb
SDP
Supplier:  ABclonal Technology A2303120UL
Encompass_Preferred
Puromycin is an aminonucleoside antibiotic derived from the Streptomyces alboniger bacterium that causes premature chain termination during translation taking place in the ribosome It has a role as a nucleoside antibiotic an antiinfective agent an antineoplastic agent a protein synthesis inhibitor an antimicrobial agent an EC 3 4 11 14 (cytosol alanyl aminopeptidase) inhibitor and an EC 3 4 14 2 (dipeptidyl-peptidase II) inhibitor It is a conjugate base of a puromycin(1 ) Puromycin is an antibiotic that prevents bacterial protein translation It is utilized as a selective agent in laboratory cell cultures Puromycin is toxic to both prokaryotic and eukaryotic cells resulting in significant cell death at appropriate doses
Catalog No. 50-000-06685