Learn More
ABclonal Technology Puromycin Rabbit mAb
SDP
Catalog No. 5000006684
Encompass_Preferred
Puromycin is an aminonucleoside antibiotic derived from the Streptomyces alboniger bacterium that causes premature chain termination during translation taking place in the ribosome It has a role as a nucleoside antibiotic an antiinfective agent an antineoplastic agent a protein synthesis inhibitor an antimicrobial agent an EC 3 4 11 14 (cytosol alanyl aminopeptidase) inhibitor and an EC 3 4 14 2 (dipeptidyl-peptidase II) inhibitor It is a conjugate base of a puromycin(1 ) Puromycin is an antibiotic that prevents bacterial protein translation It is utilized as a selective agent in laboratory cell cultures Puromycin is toxic to both prokaryotic and eukaryotic cells resulting in significant cell death at appropriate doses
Catalog No. 50-000-06684 Supplier ABclonal Technology Supplier No. A23031100UL
Puromycin is an aminonucleoside antibiotic derived from the Streptomyces alboniger bacterium that causes premature chain termination during translation taking place in the ribosome It has a role as a nucleoside antibiotic an antiinfective agent an antineoplastic agent a protein synthesis inhibitor an antimicrobial agent an EC 3 4 11 14 (cytosol alanyl aminopeptidase) inhibitor and an EC 3 4 14 2 (dipeptidyl-peptidase II) inhibitor It is a conjugate base of a puromycin(1 ) Puromycin is an antibiotic that prevents bacterial protein translation It is utilized as a selective agent in laboratory cell cultures Puromycin is toxic to both prokaryotic and eukaryotic cells resulting in significant cell death at appropriate doses