Learn More
Cayman Chemical Puromycin (hydrochloride), Purity: ≥98%, CAS: 58-58-2, 50 mg
SDP
Catalog No. 502955772
Encompass_Preferred
An inhibitor of protein synthesis; inhibits protein synthesis by 99% in vitro; an inhibitor of PSA and APN (IC50s = 9.7 and 41 µM, respectively); reduces viability of HL-60 and MOLT-4 cells (EC50s = 0.055 and 0.17 µM, respectively); active against the chloroquine-sensitive and -resistant P. falciparum strains T9-96 and K1 (IC50s = 0.024 and 0.023 µM, respectively); has been used as a selective marker in cell culture systems and has been chemically modified for use in labeling or imaging newly synthesized proteins
Catalog No. 50-295-5772 Supplier Cayman Chemical Supplier No. 1388450 MG
An inhibitor of protein synthesis; inhibits protein synthesis by 99% in vitro; an inhibitor of PSA and APN (IC50s = 9.7 and 41 µM, respectively); reduces viability of HL-60 and MOLT-4 cells (EC50s = 0.055 and 0.17 µM, respectively); active against the chloroquine-sensitive and -resistant P. falciparum strains T9-96 and K1 (IC50s = 0.024 and 0.023 µM, respectively); has been used as a selective marker in cell culture systems and has been chemically modified for use in labeling or imaging newly synthesized proteins