Learn More
Microsolv Technology Corporation Caps, Screw Top GL45 Threads, Purple, Solid without a Liner. 10/PK.
Catalog No. 502270188
Encompass_Preferred
These Caps are made of Polypropylene and have GL45 threads making them a good choice for Mobile Phase Reservoir Bottles when the Bottle is empty to prevent dust and dirt from entering it.
This item is not returnable. View return policy
Catalog No. 50-227-0188 Supplier Microsolv Technology Corporation Supplier No. 58807P45
These Caps are made of Polypropylene and have GL45 threads making them a good choice for Mobile Phase Reservoir Bottles when the Bottle is empty to prevent dust and dirt from entering it.