Learn More
Neta Scientific Agilent Purple screw caps PTFE/Silicone septa
SDP
Catalog No. 500109962
Encompass_Preferred
This item is manufactured by Agilent
Agilent Cap, screw, purple, PTFE/silicone septa, 100/pk. Cap size: 12 mm - 2 ml Screw Top Vials & Caps
UNSPSC Code  41121806
- 58
Catalog No. 50-010-9962 Supplier Neta Scientific Supplier No. 50404681
This item is manufactured by Agilent
Agilent Cap, screw, purple, PTFE/silicone septa, 100/pk. Cap size: 12 mm - 2 ml Screw Top Vials & Caps
UNSPSC Code  41121806
- 58