Learn More
Cayman Chemical Purpurogallin, Purity: ≥95%, CAS: 569-77-7, 50 mg
SDP
Catalog No. 502988840
Encompass_Preferred
A phenol; active against a range of bacteria (MICs = 11-110 µg/ml) and P. falciparum strain FCB1 clone NC-1 (IC50 = 55 µM); scavenges DPPH radicals in a cell-free assay and reduces hydrogen peroxide and radiation-induced production of ROS in HaCaT keratinocytes at 2, 5, and 10 µM; inhibits the activity of EGFR, GST, prolyl endopeptidase, and glyoxalase I (IC50s = 27.5, 8, 16, and 50 µM, respectively), as well as COMT (Ki = 0.074 µM), in cell-free assays
Catalog No. 50-298-8840 Supplier Cayman Chemical Supplier No. 2968950 MG
A phenol; active against a range of bacteria (MICs = 11-110 µg/ml) and P. falciparum strain FCB1 clone NC-1 (IC50 = 55 µM); scavenges DPPH radicals in a cell-free assay and reduces hydrogen peroxide and radiation-induced production of ROS in HaCaT keratinocytes at 2, 5, and 10 µM; inhibits the activity of EGFR, GST, prolyl endopeptidase, and glyoxalase I (IC50s = 27.5, 8, 16, and 50 µM, respectively), as well as COMT (Ki = 0.074 µM), in cell-free assays