Learn More
AdipoGen Purpuromycin
SDP
Supplier:  AdipoGen AGCN20317M005
Encompass_Preferred
Chemical. CAS 53969-01-0. Formula C26H18O13. MW 538.4. Isolated from Actinoplanes sp. Rubromycin antibiotic. Antibacterial, antifungal and anti-protozoal agent. Active against Gram-positive bacteria, Gram-negative bacteria and fungi. Telomerase inhibitor IC50=3µM. Protein synthesis inhibitor. Binds tRNAs with high affinity and specifically inhibits aminoacyl-tRNA formation consequent inhibition of protein synthesis in these microorganisms. Inhibits of all tRNA synthetases. Effectively inhibits bacterial translation and translation in eukaryotic cells. Modest inhibitor of retroviral reverse transcriptase.
Catalog No. 50-256-2606