Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PURUS™ Synthetic Cleanroom Paper
GREENER_CHOICE

Catalog No. 19125999 Shop All Purus International Inc. Products
Change view
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Page Format:
3 Hole Punch
Plain
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Page Format
19-125-999 Yellow Plain
19-125-909 Green Plain
19-125-910 Pink Plain
19-125-906 White Plain
19-125-911 Blue Plain
19-125-946 Blue 3 Hole Punch
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-125-999 Supplier PURUS™ Supplier No. LCIY 2012C
Add to Cart
Add to Cart

Cleanroom paper that excels in Tensile Strength. For writing, printing and photocopying in cleanroom environments, PURUS™ Cleanroom Paper provides excellent printability and heat toner adhesion and is recyclable.

  • Features high tensile strength and excellent heat resistance for enhanced copier and printing efficiency
  • Proprietary binding technology results in very low levels of particles, nonvolatile residue (NVR) and ion extractables
  • Unique precision cutting technology guarantees consistent sizing and cleanliness
  • Produced in ISO 4 (Class 10) cleanroom
  • Recyclable
  • Suitable for use in ISO 4 to 5 (Class 10 to Class 1000) cleanrooms

Compliance

Suitable for use in ISO 4 to 6 (Class 10 to Class 1000)

Specifications

Material Synthetic
Ruled No
Color Yellow
Dimensions (L x W) 8.5 x 11 in.
Autoclavable Autoclavable
For Use With (Application) For Class 10 to Class 1000 cleanrooms, writing, printing and photocopying in clean environments
Page Format Plain
Plain paper Yes
Product Type Cleanroom Paper
Quantity 250 Sheets/Ream, 10 Reams/Cs.
Weight (English) 22 lb.
Weight (Metric) 9.97 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.