Learn More
Apexbio Technology LLC Purvalanol A 212844-53-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000088617
Encompass_Preferred
Purvalanol A (CAS 212844-53-6) is a potent and selective inhibitor of cyclin-dependent kinases (CDKs) exhibiting IC50 values of 4 nM for cdc2-cyclin B 35 nM for CDK2-cyclin E 70 nM for CDK2-cyclin A 75 nM for CDK5-p35 and 850 nM for CDK4-cyclin D1 By targeting CDKs Purvalanol A disrupts cell cycle regulation transcription and neuronal differentiation processes In cellular studies it decreases levels of anti-apoptotic proteins and induces apoptosis partly through inhibition of JAK2/STAT3 signaling and RNA polymerase II phosphorylation Additionally Purvalanol A modulates polyamine catabolic pathways and affects cell proliferation in neural tissues supporting its utility in cell cycle apoptosis and neurobiology research
Catalog No. 50-000-88617 Supplier Apexbio Technology LLC Supplier No. B49905.1
Purvalanol A (CAS 212844-53-6) is a potent and selective inhibitor of cyclin-dependent kinases (CDKs) exhibiting IC50 values of 4 nM for cdc2-cyclin B 35 nM for CDK2-cyclin E 70 nM for CDK2-cyclin A 75 nM for CDK5-p35 and 850 nM for CDK4-cyclin D1 By targeting CDKs Purvalanol A disrupts cell cycle regulation transcription and neuronal differentiation processes In cellular studies it decreases levels of anti-apoptotic proteins and induces apoptosis partly through inhibition of JAK2/STAT3 signaling and RNA polymerase II phosphorylation Additionally Purvalanol A modulates polyamine catabolic pathways and affects cell proliferation in neural tissues supporting its utility in cell cycle apoptosis and neurobiology research