Learn More
Medchemexpress LLC HY-18299A 25mg Medchemexpress, Purvalanol A CAS:212844-53-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY18299A25MG
Encompass_Preferred
Medchemexpress, HY-18299A 25mg Purvalanol A CAS:212844-53-6 Purvalanol A is a potent CDK inhibitor, which inhibits cdc2-cyclin B, cdk2-cyclin A, cdk2-cyclin E, cdk4-cyclin D1, and cdk5-p35 with IC50s of 4, 70, 35, 850, 75 nM, resepctively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2033