Learn More
AdipoGen Purvalanol A (5 mg)
SDP
Catalog No. 502053870
Encompass_Preferred
Chemical. CAS: 212844-53-6. Formula: C19H25ClN6O. MW: 388.9. Synthetic. Potent, cell permeable cyclin-dependent protein kinase (cdk) inhibitor. Inhibits human CDK1 (IC50=4nM), CDK2/cyclin A (IC50=70nM), Cdc2/cyclin B, CDK2/cyclin E (IC50=35nM), CDK4/cyclin D1 (IC50=850nM) as well as CDK5/p35 (IC50=75nM). DYRK1A inhibitor (IC50=300nM). Anticancer agent. Strong apoptotic and autophagy inducer which causes cell cycle arrest in the G1 and G2 phase in various cancer cell lines. More membrane permeable than purvalanol B. Shown to inhibit ABCB1 and ABCB2 transporters and useful in co-treatment with selected anticancer drugs.
Catalog No. 50-205-3870 Supplier AdipoGen Supplier No. AGCR12903M005
Chemical. CAS: 212844-53-6. Formula: C19H25ClN6O. MW: 388.9. Synthetic. Potent, cell permeable cyclin-dependent protein kinase (cdk) inhibitor. Inhibits human CDK1 (IC50=4nM), CDK2/cyclin A (IC50=70nM), Cdc2/cyclin B, CDK2/cyclin E (IC50=35nM), CDK4/cyclin D1 (IC50=850nM) as well as CDK5/p35 (IC50=75nM). DYRK1A inhibitor (IC50=300nM). Anticancer agent. Strong apoptotic and autophagy inducer which causes cell cycle arrest in the G1 and G2 phase in various cancer cell lines. More membrane permeable than purvalanol B. Shown to inhibit ABCB1 and ABCB2 transporters and useful in co-treatment with selected anticancer drugs.