Learn More
Medchemexpress LLC Purvalanol B (NG 95) | 212844-54-7 | 97.0% | 432.90 | 200 MG
SDP
Catalog No. 5000387693
Encompass_Preferred
Purvalanol B (NG 95) is a potent, selective, reversible, and ATP-competitive inhibitor of cyclin-dependent kinases (CDK). It demonstrates significant selectivity for CDK over other protein kinases and effectively inhibits the growth of a chloroquine-resistant strain of *P. falciparum*.
  • Potent, selective, reversible, and ATP-competitive inhibitor of CDK
  • Inhibits cdc2-cyclin B, CDK2-cyclin A, and CDK5-p35 with IC50s of 6 nM
  • Inhibits CDK2-cyclin E with an IC50 of 9 nM
  • Shows selectivity for CDK over other protein kinases (IC50 > 1000 nM)
  • Inhibits the growth of a chloroquine-resistant strain of *P. falciparum*
Catalog No. 50-003-87693 Supplier Medchemexpress LLC Supplier No. HY18299200MG
Purvalanol B (NG 95) is a potent, selective, reversible, and ATP-competitive inhibitor of cyclin-dependent kinases (CDK). It demonstrates significant selectivity for CDK over other protein kinases and effectively inhibits the growth of a chloroquine-resistant strain of *P. falciparum*.
  • Potent, selective, reversible, and ATP-competitive inhibitor of CDK
  • Inhibits cdc2-cyclin B, CDK2-cyclin A, and CDK5-p35 with IC50s of 6 nM
  • Inhibits CDK2-cyclin E with an IC50 of 9 nM
  • Shows selectivity for CDK over other protein kinases (IC50 > 1000 nM)
  • Inhibits the growth of a chloroquine-resistant strain of *P. falciparum*