Learn More
Cayman Chemical Purvalanol B, Purity: ≥95%, CAS: 212844-54-7, 10 mg
SDP
Catalog No. 502983303
Encompass_Preferred
A CDK inhibitor that most potently inhibits Cdk2/cyclin A, Cdk2/cyclin E, Cdk5/p35, and Cdc2/cyclin B (IC50s = 6, 9, 6, and 6 nM, respectively); inhibits the growth of parasites, including Plasmodium, at micromolar doses
Catalog No. 50-298-3303 Supplier Cayman Chemical Supplier No. 1911510 MG
A CDK inhibitor that most potently inhibits Cdk2/cyclin A, Cdk2/cyclin E, Cdk5/p35, and Cdc2/cyclin B (IC50s = 6, 9, 6, and 6 nM, respectively); inhibits the growth of parasites, including Plasmodium, at micromolar doses