Learn More
Aviva Systems Biology Corp Pus1 Antibody - N-terminal region (ARP37260_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37260_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pus1. It was validated on Western Blot. Pus1,pseudouridine synthase 1,MPUS1; mPus1p; A730013B20Rik; Pus1,tRNA pseudouridine synthase A, mitochondrial,tRNA pseudouridine synthase A, mitochondrial,OTTMUSP00000027965,OTTMUSP00000027966,OTTMUSP00000027967,OTTMUSP00000027968,OTTMUSP00000027969,tRNA-uridine isomerase I,tRNA pseudouridylate synthase IThe immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: A730013B20Rik, MPUS1, mPus1pApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-609