Learn More
AVANTOR MASTERFLEX (VWR LLC) Push-to-connect straight unions, polypropylene, 1/4"", 10/pk
Catalog No. NC1650265
Encompass
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop-steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth-tubing slides right out.
Catalog No. NC1650265 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3400751
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop-steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth-tubing slides right out.