Learn More
Grainger Siemens Sirius Push-in Lugs, Size S00/S0, 1/Pk.
Catalog No. 502771978
Encompass_Preferred
These push-in lugs are designed for screw mounting applications, providing a secure connection for compatible system components. They are engineered for use with various modules to ensure reliable fastening within electrical assemblies.
  • Designed for size S00 and S0 systems
  • Features a clip-on fastening method
  • Suitable for use with 3UF71.0, 3UF71.1, and 3UF71.2 modules
  • Supports operation in ambient temperatures from -20°C to +70°C
  • Certified for CE, UL, CSA, and CCC compliance
  • Suitable for maritime and railway applications
Catalog No. 50-277-1978 Supplier Grainger Supplier No. 13Y518
These push-in lugs are designed for screw mounting applications, providing a secure connection for compatible system components. They are engineered for use with various modules to ensure reliable fastening within electrical assemblies.
  • Designed for size S00 and S0 systems
  • Features a clip-on fastening method
  • Suitable for use with 3UF71.0, 3UF71.1, and 3UF71.2 modules
  • Supports operation in ambient temperatures from -20°C to +70°C
  • Certified for CE, UL, CSA, and CCC compliance
  • Suitable for maritime and railway applications