Learn More
MSC Parker 12511 1/4" Outside Diam, 1/8 NPTF, Nickel Plated Brass Push-to-Connect Tube Male Connector 300 Max psi, Tube to Male NPT Connection, Buna-N O-Ring
Catalog No. NC1608847
Encompass
Parker 12511 1/4" Outside Diam, 1/8 NPTF, Nickel Plated Brass Push-to-Connect Tube Male Connector 300 Max psi, Tube to Male NPT Connection, Buna-N O-Ring
Catalog No. NC1608847 Supplier MSC Supplier No. 84426121
Parker 12511 1/4" Outside Diam, 1/8 NPTF, Nickel Plated Brass Push-to-Connect Tube Male Connector 300 Max psi, Tube to Male NPT Connection, Buna-N O-Ring