Learn More
StatLab Push-up Box Wipe Dispenser
SDP
Catalog No. 22000086
Change view
Click to view available options
Mounting:
Freestanding
Catalog No. Mounting
22-000-086 Freestanding
1 options
Catalog No. 22-000-086 Supplier StatLab Supplier No. TWD00101E
Add to Cart
Add to Cart
The Push-Up Box has been designed to give you perfect dispensing of wipes
  • Remove the oval cutouts on the top and bottom of a Kimwipes or StatSwipes box and drop it into the Push-Up Box
  • The Push-Up Box prevents the top-most tissue from falling back into the Kimwipes or StatSwipes box, down to the very last one, each and every time
  • No wall installation necessary
  • Put a Push-Up Box wherever it works best ergonomically
  • Made of durable high impact polystyrene
  • Compatible with StatSwipe wipes SKU STLSL5511
Specifications
Product Type Wipe Dispenser
Color White
Length (Metric) 5 in.
Width (Metric) 127 mm
Height (Metric) 127 mm
Holds Wipes
Material Plastic
Mounting Freestanding
No. of Compartments 1
Product Style Open Top
Sterility Non-sterile