Learn More
Alkali Scientific Putrescine Dihydrochloride [1,4 Diaminobutane], 1 G
SDP
Catalog No. 5000661300
Encompass_Preferred
Putrescine Dihydrochloride is a high-purity diamine compound used in cell culture applications to enhance cellular growth and proliferation. It is a key component in the synthesis of polyamines, which are vital for cell division and function. The 1g quantity is ideal for small-scale laboratory experiments that require precise amounts of the compound for enhancing cell culture conditions. Putrescine is used in research on cell signaling, gene expression, and molecular biology, particularly in studies involving cancer, cell growth, and aging. It is commonly employed in laboratories focusing on cellular metabolism and the effects of polyamines in biological systems.
Catalog No. 50-006-61300 Supplier Alkali Scientific Supplier No. PUT002CP
Putrescine Dihydrochloride is a high-purity diamine compound used in cell culture applications to enhance cellular growth and proliferation. It is a key component in the synthesis of polyamines, which are vital for cell division and function. The 1g quantity is ideal for small-scale laboratory experiments that require precise amounts of the compound for enhancing cell culture conditions. Putrescine is used in research on cell signaling, gene expression, and molecular biology, particularly in studies involving cancer, cell growth, and aging. It is commonly employed in laboratories focusing on cellular metabolism and the effects of polyamines in biological systems.