Learn More
Alkali Scientific Putrescine Dihydrochloride [1,4 Diaminobutane], 25 G
SDP
Catalog No. 5000661605
Encompass_Preferred
Putrescine Dihydrochloride is a diamine compound used in cell culture and biochemical research. It plays a critical role in the synthesis of polyamines, which are important for cell division, gene expression, and the regulation of cellular processes. This high-purity reagent is available in a 25g quantity, suitable for larger-scale experiments and prolonged studies. It is commonly used in cancer research, gene regulation studies, and experiments exploring the effects of polyamines on cell growth and differentiation. Putrescine Dihydrochloride is used in molecular biology and biotechnology applications to enhance cell culture conditions and to study cellular metabolism in both animal and plant systems.
Catalog No. 50-006-61605 Supplier Alkali Scientific Supplier No. PUT003CP
Add to Cart
Add to Cart
Putrescine Dihydrochloride is a diamine compound used in cell culture and biochemical research. It plays a critical role in the synthesis of polyamines, which are important for cell division, gene expression, and the regulation of cellular processes. This high-purity reagent is available in a 25g quantity, suitable for larger-scale experiments and prolonged studies. It is commonly used in cancer research, gene regulation studies, and experiments exploring the effects of polyamines on cell growth and differentiation. Putrescine Dihydrochloride is used in molecular biology and biotechnology applications to enhance cell culture conditions and to study cellular metabolism in both animal and plant systems.