Learn More
Research Products International Corp Putrescine Dihydrochloride [1,4-Diaminobutane], 25 Grams
SDP
Catalog No. 50213628
Encompass_Preferred
Polyamine growth regulator. Affects synthesis, activity of macromolecules, and permeability of membranes.
Catalog No. 50-213-628 Supplier Research Products International Corp Supplier No. P4104025.0
Polyamine growth regulator. Affects synthesis, activity of macromolecules, and permeability of membranes.