Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Putrescine Dihydrochloride, MP Biomedicals™

Catalog No. MP210045005 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210045005 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210045005 Supplier MP Biomedicals, Inc Supplier No. 0210045005
Add to Cart
Add to Cart

Functions as a biogenic polyamine and precursor of spermidine. Putrescine Dihydrochloride, MP Biomedicals is formed via decarboxylation of ornithine or by decarboxylation of arginine followed by hydrolysis.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-93-7
Molecular Formula C4H14Cl2N2
Molecular Weight (g/mol) 161.07
InChI Key XXWCODXIQWIHQN-UHFFFAOYSA-N
Synonym 1,4-diaminobutane dihydrochloride, putrescine dihydrochloride, tetramethylenediamine dihydrochloride, putrescine hydrochloride, 1,4-butanediamine dihydrochloride, unii-x45sur7rhy, 1,4-butanediamine, dihydrochloride, 1,4-butanediamine, hydrochloride, 1,4-diaminobutane, dihydrochloride, butane-1,4-diamine dihydrochloride
PubChem CID 9532
IUPAC Name butane-1,4-diamine;dihydrochloride
SMILES C(CCN)CN.Cl.Cl

Specifications

Melting Point 280°C (decomposition)(literature)
pH 6 to 7 (1% aq. soln.)
Quantity 5 g
Solubility Information Soluble in water (10mg/mL, clear, colorless solution).
Formula Weight 161.1
Percent Purity ∼98%
Physical Form Crystalline Powder
Recommended Storage Room Temperature (15–30°C), desiccate, protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.