Learn More
Med Vet International McKesson Therapy Putty, XX-Soft, 2 oz., 169-10-0956, Each
Catalog No. 503045865
Encompass_Preferred
Therapy Putty McKesson 2X-Soft 2 oz. Great for grip strength exercises Made with high quality silicone polymer and is gluten, latex, nuts, egg and soy-free Putty is non-toxic and unscented Color-coded tan to distinguish it as the one with extra extra soft stiffness 2 ounces of putty
Catalog No. 50-304-5865 Supplier Med Vet International Supplier No. 1129153EA
Therapy Putty McKesson 2X-Soft 2 oz. Great for grip strength exercises Made with high quality silicone polymer and is gluten, latex, nuts, egg and soy-free Putty is non-toxic and unscented Color-coded tan to distinguish it as the one with extra extra soft stiffness 2 ounces of putty