Learn More
Med Vet International McKesson Therapy Putty, Blue, Firm, 1 lb., 169-10-0921, Each
Catalog No. 503046270
Encompass_Preferred
Therapy Putty McKesson Firm 1 lb Great for grip strength exercises Made with high quality silicone polymer and is gluten, latex, nuts, egg and soy-free Putty is nontoxic and unscented Color-coded blue so you quickly can distinguish this silicone polymer therapy putty as the one that is firm 1 lb of putty
Catalog No. 50-304-6270 Supplier Med Vet International Supplier No. 1129140EA
Therapy Putty McKesson Firm 1 lb Great for grip strength exercises Made with high quality silicone polymer and is gluten, latex, nuts, egg and soy-free Putty is nontoxic and unscented Color-coded blue so you quickly can distinguish this silicone polymer therapy putty as the one that is firm 1 lb of putty