Learn More
Leica Biosystems PV POLY-AP ANTI MOUSE IGG 110M
Catalog No. NC1648349
Encompass
PowerVision Poly-AP Anti-Mouse IgG is biotin-free, polymeric alkaline phosphatase (AP) conjugate, for the manual detection of tissue-bound mouse antibodies.
Catalog No. NC1648349 Supplier Leica Biosystems Supplier No. PV6110
PowerVision Poly-AP Anti-Mouse IgG is biotin-free, polymeric alkaline phosphatase (AP) conjugate, for the manual detection of tissue-bound mouse antibodies.