Learn More
Leica Biosystems PowerVision Poly-AP Anti-Rabbit IgG
Supplier:  Leica Biosystems PV6133
Encompass
PowerVision Poly-AP Anti-Rabbit IgG is a biotin-free, polymeric alkaline phosphatase (AP) conjugate, for the manual detection of tissue-bound rabbit antibodies.
Catalog No. NC2911540