Learn More
Leica Biosystems PV POLY-HRP ANTI-MSE IGG 110ML
Catalog No. NC1385428
Encompass
PowerVision Poly-HRP anti-Rabbit IHC Detection Systems are biotin-free, polymeric horseradish peroxidase (HRP) conjugated systems, for the manual detection of tissue-bound rabbit antibodies.
Catalog No. NC1385428 Supplier Leica Biosystems Supplier No. PV6114
PowerVision Poly-HRP anti-Rabbit IHC Detection Systems are biotin-free, polymeric horseradish peroxidase (HRP) conjugated systems, for the manual detection of tissue-bound rabbit antibodies.